N-(tert-Butoxycarbonyl)-D-serine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Boc-D-serine, Boc-D-Ser-OH

B2258-1G B2258-5G
TCB2258-1GEA 26.86 CAD
TCB2258-1G TCB2258-5G
N-(tert-Butoxycarbonyl)-D-serine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-D-serine

CAS Number: 6368-20-3
MDL Number: MFCD00063142
Molecular Formula: C8H15NO5
Molecular Weight: 205.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 95
Specific rotation [a]20/D: 8.5 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-serine, Boc-D-Ser-OH

Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00063142
CAS Number: 6368-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0bc200c1ef020a6674463df6ce39b5abbacf054&vid=d0bc200c1ef020a6674463df6ce39b5abbacf054&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR