Methyl-4-bromo-3-methylbenzoate ≥97.0%

Supplier: TCI America

B2256-25G B2256-5G
TCB2256-25GEA 208.56 CAD
TCB2256-25G TCB2256-5G
Methyl-4-bromo-3-methylbenzoate ≥97.0%
Methyl 4-bromo-3-methylbenzoate

CAS Number: 148547-19-7
MDL Number: MFCD00673014
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 43


Other Product Names / Synonyms: 4-Bromo-m-toluic Acid Methyl Ester, 4-Bromo-3-methylbenzoic Acid Methyl Ester, Methyl 4-Bromo-m-toluate

Formula: C₉H₉BrO₂
Melting Pt: 43 °C
MDL Number: MFCD00673014
CAS Number: 148547-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12499cc98b3ab99ad032c45911779443a71fa9eb&vid=12499cc98b3ab99ad032c45911779443a71fa9eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR