Ethyl-5-bromo-2-chlorobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 5-bromo-2-chlorobenzoate

B2255-25G B2255-5G
TCB2255-25GEA 153.26 CAD
TCB2255-25G TCB2255-5G
Ethyl-5-bromo-2-chlorobenzoate ≥98.0%
Ethyl 5-bromo-2-chlorobenzoate

CAS Number: 76008-73-6
MDL Number: MFCD00013635
Molecular Formula: C9H8BrClO2
Molecular Weight: 263.52
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 288
Flash Point (°C): 110
Specific Gravity (20/20): 1.55


Other Product Names / Synonyms: 5-Bromo-2-chlorobenzoic Acid Ethyl Ester

Formula: C₉H₈BrClO₂
MW: 263.52 g/mol
Boiling Pt: 288 °C
Density: 1.55 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013635
CAS Number: 76008-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c92ebab969f0540c0dc012b07b5efe351e866d6&vid=1c92ebab969f0540c0dc012b07b5efe351e866d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR