Bis(neopentyl glycolato)diboron ≥98.0%

Supplier: TCI America

B2254-1G B2254-25G B2254-5G
TCB2254-1GEA 57.88 CAD
TCB2254-1G TCB2254-25G TCB2254-5G
Bis(neopentyl glycolato)diboron ≥98.0%
Bis(neopentyl glycolato)diboron

CAS Number: 201733-56-4
MDL Number: MFCD02093062
Molecular Formula: C10H20B2O4
Molecular Weight: 225.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 192
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Suzuki-Miyaura Cross-coupling Reaction, Boration
Other Product Names / Synonyms: 5,5,5',5'-Tetramethyl-2,2'-bi-1,3,2-dioxaborinane

Formula: C10H20B2O4
Melting Pt: 192 °C
MDL Number: MFCD02093062
CAS Number: 201733-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=945940f566c11a1f502be2c5b5e5fd7872dff60b&vid=945940f566c11a1f502be2c5b5e5fd7872dff60b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR