4-tert-Butylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid
B2251-25G
B2251-5G
TCB2251-25GEA
146.94
CAD
TCB2251-25G
TCB2251-5G
4-tert-Butylphenylboronic acid (contains varying amounts of Anhydride)
4-tert-Butylphenylboronic acid
CAS Number: 123324-71-0
MDL Number: MFCD01009697
Molecular Formula: C10H15BO2
Molecular Weight: 178.04
Form: Crystal
Color: White
Melting point (°C): 184
Other Product Names / Synonyms: 4-tert-Butylbenzeneboronic Acid
Formula:
C₁₀H₁₅BO₂ MW: 178.04 g/mol Melting Pt: 184 °C |
MDL Number:
MFCD01009697 CAS Number: 123324-71-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...