Benzyl (S)-(+)-glycidyl ether ≥98.0%

Supplier: TCI America

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

B2239-5G B2239-1G B2239-25G
TCB2239-5GEA 45.82 CAD
TCB2239-5G TCB2239-1G TCB2239-25G
Benzyl (S)-(+)-glycidyl ether ≥98.0%
Benzyl (S)-(+)-glycidyl ether

CAS Number: 16495-13-9
MDL Number: MFCD00054428
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 109
Specific Gravity (20/20): 1.08
Specific rotation [a]20/D: 16 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-1-Benzyloxy-2,3-epoxypropane, (S)-(+)-2-(Benzyloxymethyl)oxirane

Formula: C₁₀H₁₂O₂
Boiling Pt: 130 °C
Density: 1.08 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00054428
CAS Number: 16495-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde34d7980526ddbc36c0fbf26a403503f3ebf4d&vid=bde34d7980526ddbc36c0fbf26a403503f3ebf4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR