Benzyl (R)-(-)-glycidyl ether ≥98.0%

Supplier: TCI America

Synonyms: (R)-2-((Benzyloxy)methyl)oxirane

B2238-1G B2238-5G B2238-25G
TCB2238-1GEA 33.18 CAD
TCB2238-1G TCB2238-5G TCB2238-25G
Benzyl (R)-(-)-glycidyl ether ≥98.0%
Benzyl (R)-(-)-glycidyl ether

CAS Number: 14618-80-5
MDL Number: MFCD00068409
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 109
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -15 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-1-Benzyloxy-2,3-epoxypropane, (R)-(-)-2-(Benzyloxymethyl)oxirane

Formula: C₁₀H₁₂O₂
Boiling Pt: 130 °C
Density: 1.07 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00068409
CAS Number: 14618-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6677e4a55aa388780d491d5600fbf2e8e9a0644&vid=f6677e4a55aa388780d491d5600fbf2e8e9a0644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR