5-Bromopyrimidine ≥98.0%

Supplier: TCI America

B2229-25G B2229-5G
TCB2229-25GEA 456.62 CAD
TCB2229-25G TCB2229-5G
5-Bromopyrimidine ≥98.0%
5-Bromopyrimidine

CAS Number: 4595-59-9
MDL Number: MFCD00006117
Molecular Formula: C4H3BrN2
Molecular Weight: 158.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 74

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C
Melting Pt: 74 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8738b399a64a6c604ca27362b0873becc7a8f41&vid=d8738b399a64a6c604ca27362b0873becc7a8f41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR