1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥96.0%

Supplier: TCI America

Synonyms: Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

B2225-1G B2225-25G B2225-5G
TCB2225-1GEA 45.82 CAD
TCB2225-1G TCB2225-25G TCB2225-5G
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥96.0%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride

CAS Number: 14647-23-5
MDL Number: MFCD00013313
Molecular Formula: C26H24Cl2NiP2
Molecular Weight: 528.02
Purity/Analysis Method: >96.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Precise Polymerization Reagents, Polymerization Catalysts
Other Product Names / Synonyms: [1,2-Bis(diphenylphosphino)ethane]dichloronickel(II), Dichloro[1,2-bis(diphenylphosphino)ethane]nickel(II)

Formula: C₂₆H₂₄Cl₂NiP₂
MW: 528.016 g/mol
MDL Number: MFCD00013313
CAS Number: 14647-23-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02d715710cfd5d8ef019031920a9c4c4aa2bc406&vid=02d715710cfd5d8ef019031920a9c4c4aa2bc406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR