5-Bromo-2-hydroxypyridine 98,0
Supplier: TCI America
Synonyms:
5-Bromo-2-pyridinol, 3-Bromo-6-hydroxypyridine, 3-Bromo-6-pyridinol, 5-Bromopyridin-2-ol
B2207-25G
B2207-5G
TCB2207-25GEA
117.39
CAD
TCB2207-25G
TCB2207-5G
5-Bromo-2-hydroxypyridine 98,0
5-Bromo-2-hydroxypyridine
CAS Number: 13466-38-1
MDL Number: MFCD00234042
Molecular Formula: C5H4BrNO
Molecular Weight: 174.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 180
Other Product Names / Synonyms: 5-Bromo-2-pyridinol, 5-Bromo-2(1H)-pyridinone, 5-Bromo-2-pyridone
Formula:
C₅H₄BrNO MW: 174 g/mol Melting Pt: 180 °C |
MDL Number:
MFCD00234042 CAS Number: 13466-38-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...