Methyl-6-bromo-2-naphthoate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 6-bromo-2-naphthoate
B2202-25G
B2202-5G
TCB2202-25GEA
319.16
CAD
TCB2202-25G
TCB2202-5G
Methyl-6-bromo-2-naphthoate ≥98.0%
Methyl 6-bromo-2-naphthoate
CAS Number: 33626-98-1
MDL Number: MFCD00100408
Molecular Formula: C12H9BrO2
Molecular Weight: 265.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125
Other Product Names / Synonyms: 6-Bromo-2-naphthalenecarboxylic Acid Methyl Ester, 6-Bromo-2-naphthoic Acid Methyl Ester, Methyl 6-Bromo-2-naphthalenecarboxylate
Formula:
C₁₂H₉BrO₂ MW: 265.11 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD00100408 CAS Number: 33626-98-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...