1-Bromo-3-isopropoxybenzene ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromophenyl isopropyl ether
B2198-25G
B2198-5G
TCB2198-25GEA
147.1
CAD
TCB2198-25G
TCB2198-5G
1-Bromo-3-isopropoxybenzene ≥98.0%
1-Bromo-3-isopropoxybenzene
CAS Number: 131738-73-3
MDL Number: MFCD00070756
Molecular Formula: C9H11BrO
Molecular Weight: 215.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 109
Other Product Names / Synonyms: 2-(3-Bromophenoxy)propane, 3-Bromophenyl Isopropyl Ketone, 3-Isopropoxy-1-bromobenzene, Isopropyl 3-Bromophenyl Ketone
Formula:
C₉H₁₁BrO MW: 215.09 g/mol Flash Pt: 109 °C |
MDL Number:
MFCD00070756 CAS Number: 131738-73-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...