2-Bromo-4,6-dichloroaniline ≥97.0%
Supplier: TCI America
B2197-5G
B2197-25G
TCB2197-5GEA
97.49
CAD
TCB2197-5G
TCB2197-25G
2-Bromo-4,6-dichloroaniline ≥97.0%
2-Bromo-4,6-dichloroaniline
CAS Number: 697-86-9
MDL Number: MFCD00040936
Molecular Formula: C6H4BrCl2N
Molecular Weight: 240.91
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 273
Melting point (°C): 81
Formula:
C₆H₄BrCl₂N MW: 240.91 Boiling Pt: 273 Melting Pt: 81 °C |
MDL Number:
MFCD00040936 CAS Number: 697-86-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...