[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride ≥98.0%

Supplier: TCI America

Synonyms: Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)

B2192-1G B2192-5G
TCB2192-1GEA 154.84 CAD
TCB2192-1G TCB2192-5G
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride ≥98.0%
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride

CAS Number: 59831-02-6
MDL Number: MFCD03844773
Molecular Formula: C27H26Cl2P2Pd
Molecular Weight: 589.77
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)

Formula: C₂₇H₂₆Cl₂P₂Pd
MW: 589.77 g/mol
MDL Number: MFCD03844773
CAS Number: 59831-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8678fc707c0ef1d3fe04e19f6da4269b4150f2c2&vid=8678fc707c0ef1d3fe04e19f6da4269b4150f2c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR