1-Bromo-2,4-dichlorobenzene ≥98.0%
Supplier: TCI America
Synonyms:
2,4-Dichlorobromobenzene
B2187-5G
B2187-25G
TCB2187-5GEA
131.14
CAD
TCB2187-5G
TCB2187-25G
1-Bromo-2,4-dichlorobenzene ≥98.0%
1-Bromo-2,4-dichlorobenzene
CAS Number: 1193-72-2
MDL Number: MFCD00018333
Molecular Formula: C6H3BrCl2
Molecular Weight: 225.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 235
Flash Point (°C): 98
Freezing point (°C): 24
Formula:
C₆H₃BrCl₂ MW: 225.9 g/mol Boiling Pt: 235 °C Flash Pt: 98 °C |
MDL Number:
MFCD00018333 CAS Number: 1193-72-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...