5-Benzoylpentanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2182-25G B2182-5G
TCB2182-25GEA 327.22 CAD
TCB2182-25G TCB2182-5G
5-Benzoylpentanoic acid ≥98.0% (by GC, titration analysis)
5-Benzoylpentanoic acid

CAS Number: 4144-62-1
MDL Number: MFCD00014380
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 77


Other Product Names / Synonyms: 6-Oxo-6-phenylhexanoic Acid

Formula: C₁₂H₁₄O₃
Melting Pt: 77 °C
MDL Number: MFCD00014380
CAS Number: 4144-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42086ae1c8af283e6d744177e9adb96b24feaf73&vid=42086ae1c8af283e6d744177e9adb96b24feaf73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR