Boc-D-Alanine ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

B2175-25G B2175-5G
TCB2175-25GEA 82 CAD
TCB2175-25G TCB2175-5G
Boc-D-Alanine ≥98.0%
Boc-D-Alanine

CAS Number: 7764-95-6
MDL Number: MFCD00063123
Molecular Formula: C8H15NO4
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 84
Specific rotation [a]20/D: 26 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

Formula: C₈H₁₅NO₄
Melting Pt: 84 °C
MDL Number: MFCD00063123
CAS Number: 7764-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aee2de3dc541a1996f74fe90aa97fb197f68f4f1&vid=aee2de3dc541a1996f74fe90aa97fb197f68f4f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR