5-Bromo-2-hydroxybenzyl alcohol ≥98.0%
Supplier: TCI America
B2164-5G
B2164-1G
TCB2164-5GEA
110.6
CAD
TCB2164-5G
TCB2164-1G
5-Bromo-2-hydroxybenzyl alcohol ≥98.0%
5-Bromo-2-hydroxybenzyl alcohol
CAS Number: 2316-64-5
MDL Number: MFCD00004618
Molecular Formula: C7H7BrO2
Molecular Weight: 203.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108
Other Product Names / Synonyms: 4-Bromo-2-hydroxymethylphenol, 2-Hydroxymethyl-4-bromophenol
Formula:
C₇H₇BrO₂ Melting Pt: 108 °C |
MDL Number:
MFCD00004618 CAS Number: 2316-64-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...