2-Bromo-5-fluoroaniline ≥98.0%

Supplier: TCI America

B2163-25G B2163-5G
TCB2163-25GEA 246.32 CAD
TCB2163-25G TCB2163-5G
2-Bromo-5-fluoroaniline ≥98.0%
2-Bromo-5-fluoroaniline

CAS Number: 1003-99-2
MDL Number: MFCD00070750
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₆H₅BrFN
Boiling Pt: 154 °C
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD00070750
CAS Number: 1003-99-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab1b7a83e56396f6f75da2470c70e10f8901f2ff&vid=ab1b7a83e56396f6f75da2470c70e10f8901f2ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR