1-tert-Butyl-4-nitrobenzene ≥97.0%
Supplier: TCI America
B2149-25G
B2149-5G
TCB2149-25GEA
207.33
CAD
TCB2149-25G
TCB2149-5G
1-tert-Butyl-4-nitrobenzene ≥97.0%
1-tert-Butyl-4-nitrobenzene
CAS Number: 3282-56-2
MDL Number: MFCD00041253
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 267
Specific Gravity (20/20): 1.07
Formula:
C₁₀H₁₃NO₂ Boiling Pt: 267 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00041253 CAS Number: 3282-56-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...