4-Bromo-3,5-xylenol ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene
B2146-25G
B2146-5G
TCB2146-25GEA
170.64
CAD
TCB2146-25G
TCB2146-5G
4-Bromo-3,5-xylenol ≥98.0%
4-Bromo-3,5-xylenol
CAS Number: 7463-51-6
MDL Number: MFCD00002315
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 115
Other Product Names / Synonyms: 4-Bromo-3,5-xylenol
Formula:
C₈H₉BrO MW: 201.06 Boiling Pt: 259 Melting Pt: 116 |
MDL Number:
MFCD00002315 CAS Number: 7463-51-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...