4-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Benzyloxybenzeneboronic acid, (4-(Benzyloxy)phenyl)boronic acid

B2145-5G B2145-1G
TCB2145-005GEA 148.52 CAD
TCB2145-005G TCB2145-1G
4-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Benzyloxy)phenylboronic acid

CAS Number: 146631-00-7
MDL Number: MFCD01075705
Molecular Formula: C13H13BO3
Molecular Weight: 228.05
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-Benzyloxybenzeneboronic Acid

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
MDL Number: MFCD01075705
CAS Number: 146631-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR