3-Bromo-4-fluorobenzaldehyde ≥97.0%

Supplier: TCI America

B2143-25G B2143-5G
TCB2143-25GEA 178.54 CAD
TCB2143-25G TCB2143-5G
3-Bromo-4-fluorobenzaldehyde ≥97.0%
3-Bromo-4-fluorobenzaldehyde

CAS Number: 77771-02-9
MDL Number: MFCD00042186
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >97.0% (GC)
Form: Lump
Boiling point (°C): 139
Melting point (°C): 33
Flash Point (°C): 110

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 139 °C
Melting Pt: 33 °C
Flash Pt: 110 °C
MDL Number: MFCD00042186
CAS Number: 77771-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb88bb42e3f54b52a1458772803eaa259d1307ae&vid=84ef78ce9797fcff84a9fb4128c6c9ae225ad998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR