5,5'-Dimethyl-2,2'-bipyridyl ≥98.0%

Supplier: TCI America

Synonyms: 5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline

B2138-25G B2138-5G B2138-1G
TCB2138-25GEA 556.16 CAD
TCB2138-25G TCB2138-5G TCB2138-001G
5,5'-Dimethyl-2,2'-bipyridyl ≥98.0%
5,5'-Dimethyl-2,2'-bipyridyl

CAS Number: 1762-34-1
MDL Number: MFCD01740554
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6,6'-Bi-3-picoline, 5,5'-Dimethyl-2,2'-dipyridyl, 6,6'-Di-3-picoline, 6,6'-Di-3-picolyl

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 115 °C
MDL Number: MFCD01740554
CAS Number: 1762-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=577baf144dd77b60f629c93230247e9459ce83e6&vid=577baf144dd77b60f629c93230247e9459ce83e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR