3-Bromobenzaldehyde ethylene acetal ≥97.0%
Supplier: TCI America
Synonyms:
2-(3-Bromophenyl)-1,3-dioxolane
B2098-5G
B2098-25G
TCB2098-005GEA
82.16
CAD
TCB2098-005G
TCB2098-025G
3-Bromobenzaldehyde ethylene acetal ≥97.0%
3-Bromobenzaldehyde ethylene acetal
CAS Number: 17789-14-9
MDL Number: MFCD00003209
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.52
Other Product Names / Synonyms: 3-Bromobenzaldehyde Ethylene Acetal
Formula:
C₉H₉BrO₂ Density: 1.52 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00003209 CAS Number: 17789-14-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...