2-Bromobenzaldehyde ethylene acetal ≥97.0%

Supplier: TCI America

B2084-5G B2084-25G
TCB2084-005GEA 55.3 CAD
TCB2084-005G TCB2084-025G
2-Bromobenzaldehyde ethylene acetal ≥97.0%
2-Bromobenzaldehyde ethylene acetal

CAS Number: 34824-58-3
MDL Number: MFCD00155124
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 121
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 2-Bromobenzaldehyde Ethylene Acetal

Formula: C₉H₉BrO₂
Density: 1.53 (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00155124
CAS Number: 34824-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b462aa09c3bb7665d6a255409a768a9122b778ec&vid=b462aa09c3bb7665d6a255409a768a9122b778ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR