[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex ≥98.0%
Supplier: TCI America
Synonyms:
Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
CAS Number: 95464-05-4
MDL Number: MFCD00015757
Molecular Formula: C34H28Cl2FeP2Pd
Molecular Weight: 816.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 268
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
Formula:
C₃₅H₃₀Cl₄FeP₂Pd MW: 816.64 g/mol Melting Pt: 268 °C |
MDL Number:
MFCD00792899 CAS Number: 95464-05-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...