Bis(tricyclohexylphosphine)palladium(II) dichloride ≥97.0%

Supplier: TCI America

Synonyms: trans-Dichlorobis(tricyclohexylphosphine)palladium(II), Dichlorobis(tricyclohexylphosphine)palladium(II)

B2055-5G B2055-1G
TCB2055-5GEA 537.2 CAD
TCB2055-5G TCB2055-001G
Bis(tricyclohexylphosphine)palladium(II) dichloride ≥97.0%
Bis(tricyclohexylphosphine)palladium(II) dichloride

CAS Number: 29934-17-6
MDL Number: MFCD00191830
Molecular Formula: C36H66Cl2P2Pd
Molecular Weight: 738.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction
Other Product Names / Synonyms: Dichlorobis(tricyclohexylphosphine)palladium(II)

Formula: C₃₆H₆₆Cl₂P₂Pd
MW: 738.19 g/mol
MDL Number: MFCD00191830
CAS Number: 29934-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec25af607648680a1f91d4f8423e2fe459ae3b87&vid=ec25af607648680a1f91d4f8423e2fe459ae3b87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR