4-Bromocumene ≥97.0%

Supplier: TCI America

Synonyms: 4-Bromoisopropylbenzene, 1-Bromo-4-isopropylbenzene

B2045-25G B2045-5G
TCB2045-25GEA 243.32 CAD
TCB2045-25G TCB2045-5G
4-Bromocumene ≥97.0%
4-Bromocumene

CAS Number: 586-61-8
MDL Number: MFCD00039159
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Melting point (°C): -22
Flash Point (°C): 94
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 1-Bromo-4-isopropylbenzene, 4-Isopropylbromobenzene

Formula: C₉H₁₁Br
Boiling Pt: 219 °C
Melting Pt: –22 °C
Density: 1.28 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00039159
CAS Number: 586-61-8
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcb97ec95fea45ef87af61c37b7bcf2c89e78922&vid=bcb97ec95fea45ef87af61c37b7bcf2c89e78922&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR