[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride ≥96.0%

Supplier: TCI America

Synonyms: 1,4-Butylenebis(diphenylphosphine)palladium dichloride, Dichloro[1,4-bis(diphenylphosphino)butane]palladium(II)

B2031-1G B2031-5G
TCB2031-1GEA 181.73 CAD
TCB2031-1G TCB2031-5G
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride ≥96.0%
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride

CAS Number: 29964-62-3
MDL Number: MFCD02093437
Molecular Formula: C28H28Cl2P2Pd
Molecular Weight: 603.80
Purity/Analysis Method: >96.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Dichloro[1,4-bis(diphenylphosphino)butane]palladium(II)

Formula: C₂₈H₂₈Cl₂P₂Pd
MW: 603.80 g/mol
MDL Number: MFCD02093437
CAS Number: 29964-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f25e83dc4a0044b0fbc01ee16bd58295dd035bb&vid=5f25e83dc4a0044b0fbc01ee16bd58295dd035bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR