Bis(tri-o-tolylphosphine)palladium(II) dichloride ≥90.0%

Supplier: TCI America

Synonyms: trans-Dichlorobis(tri-o-tolylphosphine)palladium(II), Bis[tri(o-tolyl)phosphine]palladium(II) chloride, Dichlorobis(tri-o-tolylphosphine)palladium(II), Dichlorobis(tri-o-tolylphsophine)palladium

B2026-1G B2026-5G
TCB2026-001GEA 113.76 CAD
TCB2026-001G TCB2026-5G
Bis(tri-o-tolylphosphine)palladium(II) dichloride ≥90.0%
Bis(tri-o-tolylphosphine)palladium(II) dichloride

CAS Number: 40691-33-6
MDL Number: MFCD00274659
Molecular Formula: C42H42Cl2P2Pd
Molecular Weight: 786.07
Purity/Analysis Method: >90.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Dichlorobis(tri-o-tolylphosphine)palladium(II)

Formula: C₄₂H₄₂Cl₂P₂Pd
MW: 786.07 g/mol
MDL Number: MFCD00274659
CAS Number: 40691-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=793c07691681ca4b4c97ba016b20c8e5ca6bf332&vid=793c07691681ca4b4c97ba016b20c8e5ca6bf332&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR