Benzyl-4-bromophenyl ketone ≥97.0%

Supplier: TCI America

Synonyms: 1-(4-Bromophenyl)-2-phenylethanone

B2020-5G B2020-1G
TCB2020-5GEA 349.18 CAD
TCB2020-5G TCB2020-001G
Benzyl-4-bromophenyl ketone ≥97.0%
Benzyl 4-bromophenyl ketone

CAS Number: 2001-29-8
MDL Number: MFCD00016331
Molecular Formula: C14H11BrO
Molecular Weight: 275.15
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 112

Formula: C₁₄H₁₁BrO
MW: 275.14 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00016331
CAS Number: 2001-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a101b9b163499d5d0c3224679a88ef5a44d175f3&vid=a101b9b163499d5d0c3224679a88ef5a44d175f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR