2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde, 1,4-Benzodioxan-6-carboxaldehyde, 2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde
CAS Number: 29668-44-8
MDL Number: MFCD00010092
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 299
Melting point (°C): 52
Flash Point (°C): 110
Product Categories: Synthetic Organic Chemistry, Aldehydes (Multi-component Reaction)
Other Product Names / Synonyms: 1,4-Benzodioxane-6-carboxaldehyde, 6-Formyl-1,4-benzodioxane
Formula:
C₉H₈O₃ MW: 164.16 g/mol Boiling Pt: 299 °C Melting Pt: 52 °C Flash Pt: 110 °C |
MDL Number:
MFCD00010092 CAS Number: 29668-44-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...