[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98.0%

Supplier: TCI America

Synonyms: [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II), Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)

B2016-5G B2016-1G
TCB2016-005GEA 328.96 CAD
TCB2016-005G TCB2016-001G
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98.0%
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride

CAS Number: 19978-61-1
MDL Number: MFCD00015702
Molecular Formula: C26H24Cl2P2Pd
Molecular Weight: 575.75
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II), Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)

Formula: C₂₆H₂₄Cl₂P₂Pd
MW: 575.75 g/mol
MDL Number: MFCD00015702
CAS Number: 19978-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83837996cfdc11b0d413aa3970c34b655297e99f&vid=83837996cfdc11b0d413aa3970c34b655297e99f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR