3-Bromo-2-chlorothiophene ≥97.0%
Supplier: TCI America
B2013-25G
B2013-5G
TCB2013-25GEA
167.01
CAD
TCB2013-25G
TCB2013-5G
3-Bromo-2-chlorothiophene ≥97.0%
3-Bromo-2-chlorothiophene
CAS Number: 40032-73-3
MDL Number: MFCD00043883
Molecular Formula: C4H2BrClS
Molecular Weight: 197.47
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Flash Point (°C): 102
Specific Gravity (20/20): 1.84
Formula:
C₄H₂BrClS Boiling Pt: 73 °C Density: 1.84 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00043883 CAS Number: 40032-73-3 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...