Ethyl-2-benzylacetoacetate ≥97.0%

Supplier: TCI America

B2008-25G B2008-5G
TCB2008-25GEA 304.94 CAD
TCB2008-25G TCB2008-005G
Ethyl-2-benzylacetoacetate ≥97.0%
Ethyl-2-benzylacetoacetate

CAS Number: 620-79-1
MDL Number: MFCD00009156
Molecular Formula: C13H16O3
Molecular Weight: 220.27
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 284
Flash Point (°C): 109
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 2-Acetyl-3-phenylpropionic Acid Ethyl Ester, Ethyl 2-Acetyl-3-phenylpropionate, 2-Benzylacetoacetic Acid Ethyl Ester

Formula: C₁₃H₁₆O₃
Boiling Pt: 284 °C
Density: 1.07 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00009156
CAS Number: 620-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f157022ff621589862bbed9d59afb3e867589a3a&vid=f157022ff621589862bbed9d59afb3e867589a3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR