Boc-L-tyrosine methyl ester ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B2005-5G
B2005-25G
TCB2005-005GEA
95.91
CAD
TCB2005-005G
TCB2005-025G
Boc-L-tyrosine methyl ester ≥98.0% (by HPLC, total nitrogen)
Boc-L-tyrosine methyl ester
CAS Number: 4326-36-7
MDL Number: MFCD00191181
Molecular Formula: C15H21NO5
Molecular Weight: 295.34
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 106
Specific rotation [a]20/D: 50 deg (C=1, CHCl3)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-tyrosine Methyl Ester, Boc-Tyr-OMe
Formula:
C15H21NO5 Melting Pt: 106 °C |
MDL Number:
MFCD00191181 CAS Number: 4326-36-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...