Benzophenone dimethyl acetal ≥98.0%

Supplier: TCI America

B1999-5G B1999-25G
TCB1999-005GEA 105.86 CAD
TCB1999-005G TCB1999-025G
Benzophenone dimethyl acetal ≥98.0%
Benzophenone dimethyl acetal

CAS Number: 2235-01-0
MDL Number: MFCD00025900
Molecular Formula: C15H16O2
Molecular Weight: 228.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 290
Melting point (°C): 106


Other Product Names / Synonyms: Dimethoxydiphenylmethane

Formula: C₁₅H₁₆O₂
Boiling Pt: 290 °C
Melting Pt: 106 °C
MDL Number: MFCD00025900
CAS Number: 2235-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47e7137b6e55085c3980635ee769e464547a462&vid=f47e7137b6e55085c3980635ee769e464547a462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR