3-Bromobenzylamine hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B1989-5G B1989-25G
TCB1989-005GEA 94.8 CAD
TCB1989-005G TCB1989-25G
3-Bromobenzylamine hydrochloride ≥97.0% (by titrimetric analysis)
3-Bromobenzylamine hydrochloride

CAS Number: 39959-54-1
MDL Number: MFCD00012856
Molecular Formula: C7H8BrN
Molecular Weight: 222.51
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 220

Formula: C₇H₉BrClN
Melting Pt: 220 °C
MDL Number: MFCD00012856
CAS Number: 39959-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce0a5fdbccba69a525f113ed1823d9a54d839c64&vid=ce0a5fdbccba69a525f113ed1823d9a54d839c64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR