Methyl-2-amino-5-bromobenzoate ≥99.0%

Supplier: TCI America

B1988-5G B1988-25G
TCB1988-005GEA 81.05 CAD
TCB1988-005G TCB1988-025G
Methyl-2-amino-5-bromobenzoate ≥99.0%
Methyl 2-amino-5-bromobenzoate

CAS Number: 52727-57-8
MDL Number: MFCD00144761
Molecular Formula: C8H8BrNO2
Molecular Weight: 230.06
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 72


Other Product Names / Synonyms: 2-Amino-5-bromobenzoic Acid Methyl Ester, Methyl 2-Amino-5-bromobenzoate, 5-Bromoanthranilic Acid Methyl Ester

Formula: C₈H₈BrNO₂
Melting Pt: 72 °C
MDL Number: MFCD00144761
CAS Number: 52727-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f50289a357e98328a523483d73dc9780ebe72456&vid=f50289a357e98328a523483d73dc9780ebe72456&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR