Benzofuran-2-carboxylic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

B1981-1G B1981-5G
TCB1981-001GEA 50.56 CAD
TCB1981-001G TCB1981-005G
Benzofuran-2-carboxylic acid ≥99.0% (by GC, titration analysis)
Benzofuran-2-carboxylic acid

CAS Number: 496-41-3
MDL Number: MFCD00005848
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 195


Other Product Names / Synonyms: Coumarilic Acid, Coumarone-2-carboxylic Acid

Formula: C₉H₆O₃
Melting Pt: 195 °C
MDL Number: MFCD00005848
CAS Number: 496-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=940fd43fdf8cb96498190142646d78df8eb9a9ac&vid=940fd43fdf8cb96498190142646d78df8eb9a9ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR