N-Boc-D-phenylalaninol ≥97.0%

Supplier: TCI America

B1966-1G B1966-5G
TCB1966-1GEA 60.04 CAD
TCB1966-1G TCB1966-5G
N-Boc-D-phenylalaninol ≥97.0%
N-Boc-D-phenylalaninol

CAS Number: 106454-69-7
MDL Number: MFCD00216472
Molecular Formula: C14H21NO3
Molecular Weight: 251.33
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 95
Specific rotation [a]20/D: 28 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-D-phenylalaninol, (R)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, (R)-2-(Boc-amino)-3-phenyl-1-propanol

Formula: C₁₄H₂₁NO₃
Melting Pt: 95 °C
MDL Number: MFCD00216472
CAS Number: 106454-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50f4d8c1aebd45f7a3b5ec834d9c073aff17b86e&vid=50f4d8c1aebd45f7a3b5ec834d9c073aff17b86e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR