Bis(pinacolato)diboron ≥99.0%
Supplier: TCI America
B1964-5G
B1964-1G
B1964-25G
B1964-100G
TCB1964-5GEA
154.7
CAD
TCB1964-5G
TCB1964-001G
TCB1964-25G
TCB1964-100G
Bis(pinacolato)diboron ≥99.0%
Bis(pinacolato)diboron
CAS Number: 73183-34-3
MDL Number: MFCD00799570
Molecular Formula: C12H24B2O4
Molecular Weight: 253.94
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 138
Product Categories: Synthetic Organic Chemistry, Suzuki-Miyaura Cross-coupling Reaction, Boration
Other Product Names / Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi-1,3,2-dioxaborolane
Formula:
C₁₂H₂₄B₂O₄ Melting Pt: 138 °C |
MDL Number:
MFCD00799570 CAS Number: 73183-34-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...