N-Boc-L-alaninol ≥98.0%

Supplier: TCI America

Synonyms: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

B1953-1G B1953-5G
TCB1953-1GEA 42.66 CAD
TCB1953-1G TCB1953-005G
N-Boc-L-alaninol ≥98.0%
N-Boc-L-alaninol

CAS Number: 79069-13-9
MDL Number: MFCD00043121
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 60
Specific rotation [a]20/D: -8.6 deg (C=1.2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-L-alaninol, (S)-2-(tert-Butoxycarbonylamino)-1-propanol

Formula: C₈H₁₇NO₃
Melting Pt: 60 °C
MDL Number: MFCD00043121
CAS Number: 79069-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ab74cc2b367e84bb377bd52b183edd35e80a1ea&vid=6ab74cc2b367e84bb377bd52b183edd35e80a1ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR