N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine

B1938-25G B1938-5G
TCB1938-25GEA 461.36 CAD
TCB1938-25G TCB1938-005G
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥98.0% (by titrimetric analysis)
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine

CAS Number: 93102-05-7
MDL Number: MFCD00674005
Molecular Formula: C13H23NOSi
Molecular Weight: 237.42
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 66
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C

Formula: C₁₃H₂₃NOSi
MW: 237.42 g/mol
Boiling Pt: 78 °C
Density: 0.93 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00674005
CAS Number: 93102-05-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7adca3726320eea3cf60afbb8464114f05107bed&vid=7adca3726320eea3cf60afbb8464114f05107bed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR