4-Bromo-4'-tert-butylbiphenyl ≥98.0%

Supplier: TCI America

B1913-25G B1913-5G
TCB1913-025GEA 984.34 CAD
TCB1913-025G TCB1913-005G
4-Bromo-4'-tert-butylbiphenyl ≥98.0%
4-Bromo-4'-tert-butylbiphenyl

CAS Number: 162258-89-1
MDL Number: MFCD01321141
Molecular Formula: C16H17Br
Molecular Weight: 289.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143

Formula: C₁₆H₁₇Br
Melting Pt: 143 °C
MDL Number: MFCD01321141
CAS Number: 162258-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=078ff3fe8bf9591c4fb0c7170c583e09cd6d8c90&vid=078ff3fe8bf9591c4fb0c7170c583e09cd6d8c90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR