Ethyl-7-bromoheptanoate ≥97.0%
Supplier: TCI America
B1880-25G
B1880-5G
TCB1880-025GEA
1064.45
CAD
TCB1880-025G
TCB1880-005G
Ethyl-7-bromoheptanoate ≥97.0%
Ethyl-7-bromoheptanoate
CAS Number: 29823-18-5
MDL Number: MFCD00037103
Molecular Formula: C9H17BrO2
Molecular Weight: 237.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 1.22
Other Product Names / Synonyms: 7-Bromoheptanoic Acid Ethyl Ester
Formula:
C₉H₁₇BrO₂ Boiling Pt: 145 °C Density: 1.22 (20 °C) |
MDL Number:
MFCD00037103 CAS Number: 29823-18-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...