4-tert-Butylstyrene ≥90.0% stabilized
Supplier: TCI America
B1877-500ML
B1877-25ML
TCB1877-500MLEA
218.04
CAD
TCB1877-500ML
TCB1877-025ML
4-tert-Butylstyrene ≥90.0% stabilized
4-tert-Butylstyrene
(stabilized with TBC)
CAS Number: 1746-23-2
MDL Number: MFCD00065126
Molecular Formula: C12H16
Molecular Weight: 160.26
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 81
Specific Gravity (20/20): 0.88
Storage Temperature: <0°C
Formula:
C₁₂H₁₆ Density: 0.88 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00065126 CAS Number: 1746-23-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...