1,3-Bis(tosyloxy)propane ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Propane-1,3-diyl bis(4-methylbenzenesulfonate), Propane-1,3-diyl bis(4-methylbenzenesulphonate), Trimethylene Glycol Ditosylate, 1,3-Bis(p-toluenesulfonyloxy)propane, Trimethylene Glycol Bis(p-toluenesulphonate), 1,3-Bis(p-toluenesulphonyloxy)propane, Trimethylene Glycol Bis(p-toluenesulfonate), 1,3-Propanediol Ditosylate

B1875-5G B1875-25G
TCB1875-005GEA 47.4 CAD
TCB1875-005G TCB1875-025G
1,3-Bis(tosyloxy)propane ≥98.0% (by HPLC)
1,3-Bis(tosyloxy)propane

CAS Number: 5469-66-9
MDL Number: MFCD00013392
Molecular Formula: C17H20O6S2
Molecular Weight: 384.46
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 92


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,3-Bis(p-toluenesulfonyloxy)propane, 1,3-Propanediol Ditosylate, Trimethylene Glycol Bis(p-toluenesulfonate), Trimethylene Glycol Ditosylate

Formula: C₁₇H₂₀O₆S₂
MW: 384.46 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00013392
CAS Number: 5469-66-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=012e800b8b306cf77133d30254da50ee30ef0226&vid=012e800b8b306cf77133d30254da50ee30ef0226&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR