2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥96.0%

Supplier: TCI America

Synonyms: N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonyl)imide, 2-[N,N-Bis(Trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)triflimide, 2-[N,N-Bis(trifluoromethylsulphonyl)amino]-5-chloropyridine, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide), N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonyl)imide, 2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine, Comins Triflating Reagent, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulphonimide), 2-[N,N-Bis(Trifluoromethylsulfonyl)amino]-5-chloropyridine

B1865-5G B1865-25G
TCB1865-005GEA 132.72 CAD
TCB1865-005G TCB1865-25G
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine ≥96.0%
2-[N,N-Bis(trifluoromethanesulfonyl)amino]-5-chloropyridine

[Triflating Reagent]
CAS Number: 145100-51-2
MDL Number: MFCD00191833
Molecular Formula: C7H3ClF6N2O4S2
Molecular Weight: 392.67
Purity/Analysis Method: >96.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Triflation
Other Product Names / Synonyms: N-(5-Chloro-2-pyridyl)triflimide, N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonyl)imide, Comins Triflating Reagent

Formula: C₇H₃ClF₆N₂O₄S₂
MW: 392.67 g/mol
MDL Number: MFCD00191833
CAS Number: 145100-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ef81c2c8c4705c406c651445034134f9e7fb687&vid=3ef81c2c8c4705c406c651445034134f9e7fb687&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR